Learn More
Creative Biomart m7GTP Agarose 1mL
Supplier:  Creative Biomart M7GTP001A
Encompass_Preferred
Size :1 mL, Matrix :Highly cross-linked agaroseAverage particle size :45 - 165 μmLigand :m7GTPLigand concentration :5 mol m7GTP/ml gelRecommended flow rate :11.5 cm/hChemical stability :Stable to all solutions commonly used in gel filtration including 8 M urea and 6 M guanidine hydrochloride.Not stable in organic solvents!pH working range :7.5pH CIP range :4 - 9Shelf life :12 months
Catalog No. NC1337649