Learn More
Supply Solutions M9 Minimal Salts, 5X, 500mL
Supplier:  Supply Solutions SIGMA M9956500ML
Encompass_Preferred
M9 minimal media is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides, amino acids, water-soluble vitamins, and carbohydrates.
Catalog No. NC1234480