Learn More
Bioworld M9CA Medium, 500 g
Supplier:  Bioworld 306210161
Encompass_Preferred
M9CA MediumFor cultivating recombinant strains of Escherichia coli Based on M9 Minimal Salts with the addition of Casamino AcidspH: 6.8Dehydrated Appearance: light beige, homogeneous powderPrepared Medium: light to medium amberSoluble in waterContains:Bacto Casamino Acids Sodium Phosphate, Dibasic Potassium Phosphate, Monobasic Sodium Chloride Ammonium Chloride
Catalog No. 50-254-437