Learn More
Medchemexpress LLC MA-0204 100mg | 2095128-17-7 | 476.49 g/mol | C25H27F3N2O4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY114739100MG
Encompass_Preferred
MA-0204 (CAS 2095128-17-7) is a potent, highly selective, orally available peroxisome proliferator-activated receptor δ (PPARδ) modulator with low-nanomolar activity. It is supplied as a solid and as solutions in DMSO for research use only.
  • High purity: 99.31%.
  • Molecular formula C25H27F3N2O4; molecular weight 476.49 g/mol.
  • Low-nanomolar potency versus PPARδ (human EC50 ≈ 0.4 nM).
  • Available as solid and as DMSO solution for convenient experimental use.
  • Intended for research use only; not for human or clinical applications.
Catalog No. 50-002-29566