Learn More
Medchemexpress LLC MA-0204 5mg | 2095128-17-7 | 476.49 | C25H27F3N2O4 | 5 MG

Supplier: Medchemexpress LLC HY1147395MG
MA-0204 is a potent, highly selective, orally active peroxisome proliferator-activated receptor delta (PPARδ) modulator provided as a research-grade powder for preclinical studies. It demonstrates strong PPARδ activation across species and is used to probe PPARδ-mediated biology.
- Potent PPARδ modulation; human EC50 ≈ 0.4 nM, mouse EC50 ≈ 7.9 nM, rat EC50 ≈ 10 nM.
- High selectivity: >10,000-fold preference for PPARδ versus PPARα and PPARγ.
- Reported purity: 99.31% (manufacturer specification).
- Supplied as a 5 mg powder for in vitro and preclinical formulation work.
- Chemical formula C25H27F3N2O4; molecular weight 476.49 g/mol.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.