Learn More
Medchemexpress LLC Maackiain | 19908-48-6 | MFCD00270457 | 98.5% | 284.26 | C16H12O5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN038110MG
Encompass_Preferred
Maackiain is a pterocarpan phytoalexin natural product used as a research compound with reported antifungal and larvicidal activity. It is supplied in small-mass samples for biochemical and cellular studies; reported identifiers include CAS 19908-48-6, formula C16H12O5, and molecular weight 284.26.
  • Natural pterocarpan phytoalexin with bioactive properties.
  • Reported antifungal and antiparasitic (larvicidal) activity.
  • Suitable for in vitro biochemical and cellular assays.
  • Supplied as a small-mass research sample (10 mg typical package).
  • High reported purity suitable for analytical work.
Catalog No. 50-002-95983