Learn More
Bio X Cell InVivoMAb anti-mouse osteopontin (SPP1), Clone: 100D3, Size: 5 mg
Supplier:  Bio X Cell BE0372R005MG
Encompass_Preferred
InVivoMAb anti-mouse osteopontin (SPP1), Clone: 100D3, Size: 5 mg, Isotype: Mouse IgG2c, κ, Applications: in vivo OPN neutralization, in vitro OPN neutralization, ELISA
Catalog No. 50-217-2969