Learn More
Bio X Cell InVivoMAb anti-mouse osteopontin (SPP1), Clone: 100D3, Size: 50 mg
Supplier: Bio X Cell BE0372R050MG
InVivoMAb anti-mouse osteopontin (SPP1), Clone: 100D3, Size: 50 mg, Isotype: Mouse IgG2c, κ, Applications: in vivo OPN neutralization, in vitro OPN neutralization, ELISA