Learn More
Bio X Cell Anti-Mouse Osteopontin Antibody, Clone 103D6, 100 MG
Supplier: Bio X Cell BE0373R100MG
The 103D6 monoclonal antibody reacts with mouse osteopontin (OPN), also known as SPP1, a glycoprotein regulating bone development, immune responses, and oncogenesis. This blocking antibody has been shown to suppress colon tumor growth in tumor-bearing mice in vivo.
- In vivo OPN neutralization
- In vitro OPN neutralization
- ELISA application
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Endotoxin ≤1 EU/mg (≤0.001 EU/μg)
- Purity ≥95% (determined by SDS-PAGE)
- Sterility by 0.2 μm filtration
- Purified by Protein G
- Store at stock concentration at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.