Learn More
Bio X Cell InVivoMAb anti-mouse PD-L1 (B7-H1), Clone: 10F.2H11, Size: 50 mg
Supplier: Bio X Cell BE0361R050MG
InVivoMAb anti-mouse PD-L1 (B7-H1), Clone: 10F.2H11, Size: 50 mg, Isotype: Rat IgG2b, κ, Applications: in vivo blocking of PD-L1/CD80 (B7-1) interactions, in vitro blocking of PD-L1/CD80 (B7-1) interactions, ELISA, Flow cytometry