Learn More
Bio X Cell InVivoMAb Antibody, Clone 1A8, Mouse Ly6G, Rat IgG2a
Supplier: Bio X Cell BE00751R001MG
This InVivoMAb antibody, clone 1A8, targets mouse Ly6G, a GPI-anchored cell surface protein involved in cell signaling and adhesion. It is specifically designed for various research applications including in vivo neutrophil and MDSC depletion, immunofluorescence, immunohistochemistry (paraffin and frozen), and flow cytometry. The antibody exhibits specific reactivity with mouse Ly6G, with no reported cross-reactivity with Ly6C.
- Isotype: Rat IgG2a, κ
- Purity: ≥95% determined by SDS-PAGE
- Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.