Learn More
Bio X Cell Antibody, Anti-Human CSF1R (CD115), Clone 2-4A5-4, Rat IgG1
Supplier: Bio X Cell BE0347R005MG
The 2-4A5-4 monoclonal antibody targets human colony stimulating factor 1 receptor (CSF1R/CD115), a single-pass type I membrane protein. CSF1R is expressed by monocytes/macrophages, dendritic cells, and osteoclasts, regulating cellular proliferation and differentiation in the monocytic lineage.
- Reacts with human CSF1R (CD115)
- Isotype: rat IgG1, κ
- Immunogen: rat NRK cells expressing human CSF1R
- Reported applications: in vitro CSF1R neutralization, immunohistochemistry (paraffin), functional assays, flow cytometry
- Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
- Endotoxin: ≤1 EU/mg
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Purification: protein G
- Molecular weight: 150 kDa
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.