Learn More
Bio X Cell Monoclonal Antibody, 3H3 Clone, Anti-mouse 4-1BB (CD137)
Supplier: Bio X Cell BE0239R001MG
This 3H3 monoclonal antibody targets mouse 4-1BB (CD137), a TNF receptor superfamily member expressed on various immune cells. It provides costimulatory signals to T cells upon ligand binding. The antibody is an agonistic type, known to stimulate 4-1BB signaling in vivo, which is relevant for inducing T cell-mediated antitumor immunity.
- Agonistic antibody, stimulates 4-1BB signaling in vivo
- Reacts with mouse 4-1BB
- Clone 3H3
- Isotype is rat IgG2a
- Immunogen: mouse CD137 human Fc fusion protein
- Applications include in vivo and in vitro 4-1BB stimulation, and in vitro organoids/organ-on-chip
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤1EU/mg
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Molecular weight: 150 kDa
- Storage at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.