Learn More
Bio X Cell InVivoMAb anti-human CEACAM 1/3/5/6/8, Clone: 6G5J, Size: 5 mg
Supplier: Bio X Cell BE0362R005MG
InVivoMAb anti-human CEACAM 1/3/5/6/8, Clone: 6G5J, Size: 5 mg, Isotype: Mouse IgG1, κ, Applications: Western blot, ELISA, Immunofluoresence, Immunohistochemistry (paraffin), Flow cytometry