Learn More
Bio X Cell Antibody, anti-human/rat HER2 (neu), Clone 7.16.4, Mouse IgG2a
Supplier:  Bio X Cell BE0277R001MG
Encompass_Preferred
This monoclonal antibody, clone 7.16.4, targets human and rat HER2 (human epidermal growth factor receptor 2), a 185 kDa transmembrane glycoprotein. HER2 is a proto-oncoprotein often overexpressed in various tumors. This antibody inhibits HER2-overexpressing tumor growth both in vitro and in vivo, making it suitable for a range of research applications.
  • Targets human and rat HER2 (neu).
  • Clone 7.16.4 for specific binding.
  • Mouse IgG2a, κ isotype.
  • Suitable for in vivo and in vitro HER2/neu inhibition, immunoprecipitation, immunofluorescence, and flow cytometry.
  • High purity (≥95% by SDS-PAGE).
  • Sterile (0.2 μm filtration).
  • Formulated in PBS, pH 7.0, without stabilizers or preservatives.
  • Low endotoxin level (≤1EU/mg).
  • Produced in an animal-free facility.
  • For research use only.
Catalog No. 50-152-8457