Learn More
Bio X Cell InVivoMAb anti-mouse c-Kit (CD117), Clone ACK2, Rat IgG2b
Supplier:  Bio X Cell BE0293R005MG
Encompass_Preferred
The ACK2 monoclonal antibody targets mouse c-Kit, also known as CD117. c-Kit is a 145 kDa transmembrane tyrosine kinase expressed on hematopoietic progenitor cells, mast cells, and AML cells. The antibody depletes c-Kit+ cells in vivo and neutralizes c-Kit+ signaling in vitro, and is also reported to cross-react with rat c-Kit.
  • Targets mouse c-Kit (CD117)
  • Formulated in PBS, pH 7.0; contains no stabilizers or preservatives
  • Purity greater than 95%
  • Sterility: 0.2 μm filtered
  • Endotoxin level less than or equal to 1 EU/mg
  • Reported for in vivo mast cell depletion
  • Reported for in vivo c-Kit+ cell depletion
  • Reported for in vitro c-Kit neutralization
  • Reported for immunoprecipitation
  • Reported for flow cytometry
Catalog No. 50-206-5663