Learn More
Bio X Cell InVivoMAb anti-human CD71 (TfR1), Clone: B3/25, Size: 100 mg
Supplier:  Bio X Cell BE0367R100MG
Encompass
InVivoMAb anti-human CD71 (TfR1), Clone: B3/25, Size: 100 mg, Isotype: Mouse IgG1, κ, Applications: in vitro CD71 targeting, in vivo CD71 targeting, Immunofluorescence
Catalog No. 50-217-2947
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.