Learn More
Bio X Cell InVivoMAb, anti-mouse IL-12 p35, C18.2, 5 MG
Supplier: Bio X Cell BE0371R005MG
The C18.2 monoclonal antibody reacts with mouse interleukin-12 (IL-12) p35. IL-12 is a heterodimeric 70 kDa cytokine that plays a key role in the development of Th1 responses. This antibody is suitable for in vivo and in vitro applications.
- Purified by Protein G to ≥95% purity
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Endotoxin level ≤1EU/mg (≤0.001EU/μg)
- Sterile filtered to 0.2 μm
- Molecular weight: 150 kDa
- Applications: in vivo IL-12 p35 neutralization, in vitro IL-12 p35 neutralization, ELISA, immunoprecipitation
- Storage: store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.