Learn More
Bio X Cell InVivoMAb anti-human CD2, Clone: CB.219, Size: 100 mg
Supplier:  Bio X Cell BE0354R100MG
Encompass_Preferred
InVivoMAb anti-human CD2, Clone: CB.219, Size: 100 mg, Isotype: Mouse IgG2b, κ, Applications: in vivo CD2 blockade in huCD2tg mice
Catalog No. 50-217-2882