Learn More
Bio X Cell InVivoMAb anti-mouse CD69, Clone CD69.2.2, 1 MG
Supplier: Bio X Cell BE0330R001MG
The CD69.2.2 monoclonal antibody reacts with mouse CD69, an 85 kDa type II C-type lectin. It is rapidly induced upon cell activation in leukocytes and is thought to be a negative regulator of the immune response.
- Reacts with mouse CD69, an 85 kDa type II C-type lectin
- Isotype: mouse IgG1, κ
- Purified using protein A from cell culture supernatant
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Endotoxin ≤1EU/mg and purity ≥95%
- Sterility ensured by 0.2 μm filtration
- Reported for in vivo down-regulation of CD69 expression and functional assays
- Store at stock concentration at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.