Learn More
Bio X Cell recombinant Flt-3L-Ig, Flt-3L Fc-G1, Human/Human, Animal-Free
Supplier: Bio X Cell BE0342R005MG
This recombinant Flt-3L-Ig fusion protein consists of human Flt-3L fused to the Fc portion of human IgG1. Flt-3L (FMS-related Tyrosine Kinase 3 Ligand) is an endogenous protein functioning as a cytokine and growth factor, crucial for the development of conventional and plasmacytoid dendritic cells. This fusion protein is utilized to activate Flt3 signaling and induce the expansion of DC populations.
- Flt-3L Fc-G1 clone.
- Purified from an animal-free facility.
- Reacts with mouse and human.
- Formulated in PBS, pH 7.0, without stabilizers or preservatives.
- Endotoxin level of ≤1 EU/mg (≤0.001 EU/μg).
- Purity of ≥95% as determined by SDS-PAGE.
- Sterilized by 0.2 μm filtration.
- Purified by Protein A.
- Store at 4°C; do not freeze.
- Unconjugated, with conjugation services available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.