Learn More
Bio X Cell InVivoMAb anti-mouse CD24, Clone: M1/69, Size: 50 mg
Supplier:  Bio X Cell BE0360R050MG
Encompass_Preferred
InVivoMAb anti-mouse CD24, Clone: M1/69, Size: 50 mg, Isotype: Rat IgG2b, κ, Applications: in vivo administration, Immunohistochemistry (frozen), Immunohistochemistry (paraffin), Immunofluorescence, Flow cytometry
Catalog No. 50-217-2911