Learn More
Bio X Cell InVivoMAb anti-mouse FasL (CD178), Clone MFL3, Unconjugated
Supplier: Bio X Cell BE0319R050MG
The MFL3 monoclonal antibody targets mouse Fas Ligand (FasL), also known as CD178, CD95 Ligand, and TNFSF6. FasL is a 40 kDa type II transmembrane glycoprotein expressed on activated T cells, spleen, testis, and eye. This antibody is reported to block CD178/CD95 induced apoptosis, which typically induces apoptotic cell death to maintain peripheral tolerance. It is produced in an animal-free facility.
- Clone: MFL3
- Host: Armenian hamster
- Reactivity: Mouse
- Isotype: Armenian hamster IgG
- Conjugation: Unconjugated
- Reported applications: In vivo/in vitro FasL blockade, functional assay, IHC (paraffin), flow cytometry
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤1 EU/mg
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 μm filtration
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.