Learn More
Bio X Cell Antibody, CD47, Clone MIAP410, Mouse/Human/Rat, 1 MG
Supplier: Bio X Cell BE0283R001MG
The MIAP410 monoclonal antibody targets mouse CD47 (IAP), a glycosylated protein crucial for immune and angiogenic responses across various cell types. This antibody neutralizes CD47 activity in vivo and in vitro, offering value for studying its role in cellular processes and therapeutic potential.
- Reacts with mouse, human, and rat CD47
- Clone MIAP410
- Isotype: Mouse IgG1, κ
- Formulated in PBS, pH 7.0, without stabilizers or preservatives.
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Produced in an animal-free facility from cell culture supernatant.
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C, do not freeze
- Applications: in vivo/vitro CD47 blockade, immunofluorescence.
- Immunogen: Purified human placental CD47
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.