Learn More
Bio X Cell InVivoMAb, IL-27, MM27.7B1, IgG2a, Unconjugated, 1 MG
Supplier: Bio X Cell BE0326R001MG
The MM27.7B1 monoclonal antibody reacts with the p28 subunit of mouse IL-27, a member of the IL-6/IL-12 cytokine family. This clone neutralizes mouse IL-27 bioactivity in vivo and in vitro, and also binds and inhibits human IL-27.
- Purity: ≥95% (SDS-PAGE)
- Sterility: 0.2 μm filtration
- Molecular weight: 150 kDa
- Endotoxin: ≤1EU/mg (LAL assay)
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Purification: Protein A from animal-free facility
- Storage: 4°C, do not freeze
- Applications: In vivo/in vitro IL-27 p28 neutralization, flow cytometry
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.