Learn More
Bio X Cell InVivoMAb anti-human/mouse/rat amyloid-beta, Clone MOAB-2, Mouse IgG2b
Supplier: Bio X Cell BE0368R001MG
This pan-specific monoclonal antibody recognizes unaggregated, oligomeric, or fibrillar forms of Aβ42 and unaggregated Aβ40. It is selective for human Aβ42 over Aβ40, specifically detecting Aβ but not amyloid precursor protein (APP) in various homogenates, and identifies intraneuronal Aβ and amyloid plaques.
- Recognizes unaggregated, oligomeric, or fibrillar Aβ42 and unaggregated Aβ40
- Selective for human Aβ42 over Aβ40
- Detects amyloid-beta (Aβ) specifically; does not detect APP
- Identifies intraneuronal Aβ and amyloid plaques
- Reactivities: Mouse, human, rat; Isotype: Mouse IgG2b, lambda
- Formulation: PBS, pH 7.0; no stabilizers or preservatives
- Endotoxin: ≤1 EU/mg; Purity: ≥95%; Sterility: 0.2 μm filtration
- Purification: Protein A; Molecular weight: 150 kDa
- Applications: Western blot, IHC, immunofluorescence, IP, ELISA
- Storage: 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.