Learn More
Bio X Cell InVivoMAb anti-human/mouse/rat amyloid-beta, Clone: MOAB-2, Size: 5 mg
Supplier: Bio X Cell BE0368R005MG
InVivoMAb anti-human/mouse/rat amyloid-beta, Clone: MOAB-2, Size: 5 mg, Isotype: Mouse IgG2b, κ, Applications: Western blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, ELISA