Learn More
Genscript Corporation THE™ beta Actin Antibody [HRP], mAb, Mouse
Supplier:  Genscript Corporation A00730200
Encompass_Preferred
THE™ beta Actin Antibody [HRP], mAb, Mouse reacts with mouse, rabbit, chicken, human, hamster, cow, goat, fish, and pig substrates.It has not yet been tested in other species.
Catalog No. 50-272-971