Learn More
Biotium CD61 / Integrin beta-3 / Platelet Glycoprotein IIIa (Platelet Marker) (ITGB3/1713), CF405S conjugate, 0.1mg/mL
Supplier:  Biotium BNC041713500
Encompass_Preferred
This antibody reacts with human integrin beta3 (GPIIIa, vitronectin receptor beta chain). It associates with the alpha-V-chain (CD51) to form vitronectin receptor, or with the alpha-IIb-chain (CD41) to form the GpIIb/GpIIIa complex (CD41/CD61). The CD41/CD61 complex appears early in megakaryocyte maturation. The activated CD41/CD61 complex is a receptor for von Willebrand factor, soluble fibrinogen, fibronectin, vitronectin and thrombospondin. It plays a central role in platelet activation and aggregation. The CD51/CD61 is implicated in tumor metastasis and adenoviral infection. The antibody detects platelets in smears of blood and bone marrow, as well as megakaryocytes in frozen sections and cell smears. The antibody is useful for classification of megakaryoblastic leukemia.Primary antibodies are available purified, or with a selection of fluorescent CF Dyes and other labels. CF Dyes offer exceptional brightness and photostability. Note: Conjugates of blue fluorescent dye
Catalog No. 50-006-44237