Learn More
Bio X Cell InVivoMAb anti-mouse Ly6G/Ly6C, Clone NIMP-R14, Rat IgG2b, Mouse Reactivity
Supplier: Bio X Cell BE0320R005MG
The NIMP-R14 monoclonal antibody is specific for murine neutrophils, reacting strongly with mouse Ly6G and Ly6C. Ly6G is a GPI-anchored cell surface protein expressed by mature granulocytes and peripheral neutrophils, while Ly6C is a monocyte/macrophage and endothelial cell differentiation antigen. This antibody is useful for in vivo neutrophil depletion and various immunological applications.
- Clone NIMP-R14
- Reactivity: mouse
- Isotype: rat IgG2b, κ
- Immunogen: purified BALB/c mouse neutrophils
- Applications: in vivo neutrophil depletion, immunohistochemistry (paraffin), immunohistochemistry (frozen), immunofluorescence, flow cytometry
- Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
- Endotoxin: ≤1 EU/mg
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Purification: protein A
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.