Learn More
Bio X Cell Monoclonal Antibody, Clone NIMR5, Rat IgG2a, Mouse CD38, 0.2 μm Filtration
Supplier: Bio X Cell BE0317R005MG
This monoclonal antibody reacts with mouse CD38, a 42 kDa type II transmembrane glycoprotein involved in NAD homeostasis and calcium signaling. The NIMR5 clone is agonistic, inducing B cell proliferation, protecting against apoptosis, and slowing disease progression in tumor-bearing mice.
- Reacts with mouse CD38
- Clone: NIMR5
- Isotype: Rat IgG2a, kappa
- Formulation: PBS, pH 7.0, no preservatives
- Endotoxin: ≤1 EU/mg
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Produced in animal-free facility
- Purified by Protein G
- Molecular weight: 150 kDa
- Immunogen: BCL1 plasma membrane glycoproteins
- Applications: In vivo/in vitro CD38 stimulation, B cell activation, immunofluorescence, ELISA, flow cytometry
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.