Learn More
Bio X Cell InVivoMAb anti-mouse CD27, Clone: RM27-3E5, Size: 25 mg
Supplier:  Bio X Cell BE0348R025MG
Encompass_Preferred
InVivoMAb anti-mouse CD27, Clone: RM27-3E5, Size: 25 mg, Isotype: Rat IgG2a, κ, Applications: in vivo CD27 stimulation, in vitro CD27 stimulation, Immunoprecipitation, Flow cytometry
Catalog No. 50-217-2852