Learn More
Bio X Cell InVivoMAb anti-mouse CD80 (B7-1), Clone: RM80, Size: 1 mg
Supplier:  Bio X Cell BE0365R001MG
Encompass_Preferred
InVivoMAb anti-mouse CD80 (B7-1), Clone: RM80, Size: 1 mg, Isotype: Rat IgG2a, Applications: in vivo CD80 blockade, in vitro CD80 blockade, Flow cytometry
Catalog No. 50-217-2933