Learn More
Bio X Cell InVivoMAb anti-SARS-CoV-2 S protein (RBD epitope A), Clone: SARS2-01, Size: 25 mg
Supplier: Bio X Cell BE0357R025MG
InVivoMAb anti-SARS-CoV-2 S protein (RBD epitope A), Clone: SARS2-01, Size: 25 mg, Isotype: Mouse IgG1, κ, Applications: in vitro blocking of SARS-CoV-2 S protein, Flow cytometry, ELISA