Learn More
Bio X Cell InVivoMAb anti-SARS-CoV-2 S protein (RBD epitope B), Clone: SARS2-34, Size: 100 mg
Supplier:  Bio X Cell BE0359R100MG
Encompass_Preferred
InVivoMAb anti-SARS-CoV-2 S protein (RBD epitope B), Clone: SARS2-34, Size: 100 mg, Isotype: Mouse IgG1, κ, Applications: in vitro blocking of SARS-CoV-2 S protein, Flow cytometry, ELISA
Catalog No. 50-217-2907