Learn More
Bio X Cell InVivoMAb anti-human CD71 (TfR1), Clone: T56/14, Size: 100 mg
Supplier:  Bio X Cell BE0370R100MG
Encompass_Preferred
InVivoMAb anti-human CD71 (TfR1), Clone: T56/14, Size: 100 mg, Isotype: Mouse IgG1, κ, Applications: Flow cytometry, Immunoprecipitation
Catalog No. 50-217-2962