Learn More
Bio X Cell InVivoMAb anti-mouse/human PrP (prion protein), Clone: TW1, Size: 100 mg
Supplier:  Bio X Cell BE0350R100MG
Encompass_Preferred
InVivoMAb anti-mouse/human PrP (prion protein), Clone: TW1, Size: 100 mg, Isotype: Mouse IgG2a, Applications: in vivo PrP blocking, Western blot
Catalog No. 50-217-2862