Learn More
Bio X Cell InVivoMAb anti-mouse/human PrP (prion protein), Clone: TW1, Size: 5 mg
Supplier: Bio X Cell BE0350R005MG
InVivoMAb anti-mouse/human PrP (prion protein), Clone: TW1, Size: 5 mg, Isotype: Mouse IgG2a, Applications: in vivo PrP blocking, Western blot