Learn More
Medchemexpress LLC HY-44222 500mg Medchemexpress, MAC glucuronide linker-2 CAS:229977-57-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY44222500MG
Encompass_Preferred
Medchemexpress, HY-44222 500mg MAC glucuronide linker-2 CAS:229977-57-5 MAC glucuronide linker-2 is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2244