Learn More
Medchemexpress LLC Mac glucuronide phenol-linked SN-38 | 2246380-69-6 | 98.5% | 1091.06 | C50H54N6O20S | 1 MG

Supplier: Medchemexpress LLC HY1289431MG
MAC glucuronide phenol-linked SN-38 is a pH-sensitive lactone drug-linker derived from SN-38 (a DNA topoisomerase I inhibitor) intended for antibody-drug conjugate and payload-linker research. It is supplied as a solid for use in synthesis, cytotoxicity testing, and preclinical stability studies.
- pH-susceptible lactone drug-linker derived from SN-38.
- Purity reported as 98.49% by HPLC.
- Molecular formula C50H54N6O20S, molecular weight 1091.06.
- Commonly supplied in 1 mg, 5 mg, and 10 mg quantities for research use.
- Datasheet, COA, and SDS available for characterization and handling.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.