Learn More
Medchemexpress LLC HY-14456A 5mg Medchemexpress, MAC13243 CAS:1071638-38-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY14456A5MG
Encompass_Preferred
Medchemexpress, HY-14456A 5mg MAC13243 CAS:1071638-38-4 MAC13243, an antibacterial agent, is an inhibitor of bacterial lipoprotein targeting chaperone, LolA. MAC13243 is an antibacterial agent with Gram-negative selectivity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1602