Learn More
Medchemexpress LLC HY-116872 5mg Medchemexpress, MAC13772 CAS:4871-40-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1168725MG
Encompass_Preferred
Medchemexpress, HY-116872 5mg MAC13772 CAS:4871-40-3 MAC13772 is a potent inhibitor of the enzyme BioA (IC50=250 nM), the antepenultimate step in biotin biosynthesis. MAC13772 is a novel antibacterial compound. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9446