Learn More
Biocare Medical, LLC MACH 3 Mouse AP-Polymer Detection - 6.0 ml
SDP
Supplier:  Biocare Medical, LLC M3M532G
Encompass_Preferred
This biotin-free technology uses a specific probe to detect mouse monoclonal primary antibodies which is then followed by an alkaline phosphatase (AP) polymer that binds to the probe. This innovative AP-polymerization technology provides up to 5 to 20 times greater staining sensitivity compared to Envision and other polymer detection kits. It can be used manually and on automated stainers.
Catalog No. 50-828-98