Learn More
Biocare Medical, LLC MACH 3 RB AP-POLYMER 6ML
SDP
Supplier:  Biocare Medical, LLC M3R533G
Encompass_Preferred
MACH 3 Rabbit AP-Polymer is a two-step, biotin-free detection system which provides excellent specificity, sensitivity and nuclear staining for rabbit primary antibodies. It is labeled with alkaline phosphatase (AP).
Catalog No. 50-829-04