Learn More
Biocare Medical, LLC MACH 4 Universal HRP-Polymer - 6.0 ml
SDP
Supplier:  Biocare Medical, LLC M4U534G
Encompass_Preferred
The Universal HRP-Polymer detects both mouse and rabbit antibodies.This biotin-free technology uses a specific probe to detect mouse primary antibodies and is then followed by a horseradish peroxidase polymer (HRP) that binds to the probe. This innovative HRP-polymerization technology provides increased staining sensitivity for mouse monoclonal antibodies when compared to Envision and other polymer detection kits. It can be used manually and on automated stainers.
Catalog No. 50-829-10