Learn More
Essendant MACHINEMANUAL HTPRESSBG
Supplier:  Essendant IGTMS256S
Encompass_Preferred
The award-winning Model 256 manual swing-away heat press machine with 16 x20 platens swings almost a full 180 to the right providing you with unobstructed access to the lower platen while you load your garments and place your transfers The new Quick-Change platens allow you to quickly swap your lower platen with any of Insta s new accessory platens The quick-change feature also allows you to quickly rotate the lower platen in a matter of seconds to suit your applications Dimensions- 32x40x22 On/Off Indicator Light- Yes Auto Shut-Off- Yes Wattage- 1 750 W
Catalog No. 50-000-32068
Only null left
Add to Cart
Request a Quote