Learn More
Medchemexpress LLC mAChR-IN-1 hydrochloride | 119391-73-0 | MFCD01725710 | 99.6% | 524.82 | C23H26ClIN2O2 | 50MG
SDP
Supplier:  Medchemexpress LLC HY12426A50MG
Encompass_Preferred
mAChR-IN-1 hydrochloride is a research-grade muscarinic cholinergic receptor antagonist supplied as the hydrochloride salt. It exhibits high potency (IC50 = 17 nM) and is provided as a high-purity solid for in vitro pharmacology and receptor profiling.
  • High potency mAChR antagonist (IC50 = 17 nM).
  • High purity: 99.6%.
  • Hydrochloride salt form for improved stability and handling.
  • White to off-white solid suitable for in vitro assays.
  • Available in multiple pack sizes and a solution format for convenience.
  • Store sealed at 4°C; in solvent, store at -80°C for long-term stability.
Catalog No. 50-002-35368