Learn More
Selleck Chemical LLC Macitentan
SDP
Supplier:  Selleck Chemical LLC S80515MG
Encompass_Preferred
Macitentan is an orally active non-peptide dual ETA/ETB (endothelin) receptor antagonist with IC50 of 0 5 nM/391 nM
Catalog No. 50-000-00103