Learn More
Cayman Chemical MacItentn-d4 1mg
SDP
Supplier:  Cayman Chemical 285221 MG
Encompass_Preferred
An internal standard for the quantification of macitentan by GC- or LC-MS
Catalog No. 50-293-9940