Learn More
Medchemexpress LLC MAdCAM1 Human HEK2 100ug
SDP
Supplier:  Medchemexpress LLC HYP70852100UG
Encompass_Preferred
Recombinant human MAdCAM-1 produced in HEK293 cells and supplied as a lyophilized, C-terminal His-tagged protein. The protein is characterized for low endotoxin and validated functional activity by cell adhesion assays, making it suitable for receptor binding, adhesion, and migration studies. Reconstitute and store according to the lot-specific certificate of analysis for optimal stability.
  • Produced in HEK293 mammalian cells.
  • C-terminal His tag facilitates detection and purification.
  • Supplied lyophilized with lot-specific formulation details.
  • Endotoxin <1 EU/μg for cell-based applications.
  • Functional activity validated by cell adhesion assays (ED50 ~1.987 μg/mL).
  • Suitable for receptor interaction, adhesion, and migration studies.
  • Store frozen and aliquot to avoid repeated freeze-thaw cycles.
Catalog No. 50-001-96825